Tisk…
Cone supplies for PerkinElmer
Dodavatel: Agilent
8003-0582 8003-0583 8003-0585 8003-0586
HEWL8003-0582EA 1190 CZK
HEWL8003-0582 HEWL8003-0583 HEWL8003-0585 HEWL8003-0586
Cone supplies for PerkinElmer
Chromatografické potřeby Chromatography Spare Parts
These cone supplies are equivalent to the PerkinElmer ICP-MS versions for complete confidence.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the stand behind warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage