Tisk…
InfinityLab poroshell 120 PFP HPLC columns
Dodavatel: Agilent
699975-408 695975-408 695775-408 695975-308 693975-408 693975-308 699975-308 693770-408K 693970-408K 699975-308K 699975-408K 693975-408K 695775-408K 695970-408K 695975-408K
HEWL699975-408EA 22700 CZK
HEWL699975-408 HEWL695975-408 HEWL695775-408 HEWL695975-308 HEWL693975-408 HEWL693975-308 HEWL699975-308 HEWL693770-408K HEWL693970-408K HEWL699975-308K HEWL699975-408K HEWL693975-408K HEWL695775-408K HEWL695970-408K HEWL695975-408K
InfinityLab poroshell 120 PFP HPLC columns
Kolony chromatografické
Rugged, high-resolution separations up to five times faster than traditional 5 µm methods.
  • High efficiency columns for improved throughput
  • Excellent lot-to-lot reproducibility gives confidence in separation results
  • New HILIC chemistries offer a range of options for challenging polar compounds
InfinityLab Poroshell 120 columns provide exceptional efficiency and reliability, significantly boosting performance from all instruments, whether need improved HPLC performance, UHPLC performance at lower pressures or the highest UHPLC performance. ThreeMore Product Information