Tisk…
UHPLC columns, InfinityLab Poroshell 120
Dodavatel: Agilent
693975-901 699775-901 693775-901 699975-901 699975-301 695975-901 695975-301 695775-901 693975-301 699770-901 699770-901K 693770-901K 693970-301K 693970-901K 699970-901 699970-901K 695770-901 695770-901K 695970-301 695970-301K 695970-901 695970-901K 699970-301 699970-301K 650750-901K 690970-301K 820750-926 821725-926 823750-926 690970-901K 823750-940 820750-920 821725-919 820750-918 820750-919 821725-918 821725-920 823750-917 823750-918 823750-919 823750-920 820750-917 821725-917 695675-902 693675-912 693675-408 693675-901 693675-902 693675-906 695675-408 695675-901 695675-906 695675-912 821725-940 821725-941 821725-942 821725-943 821725-944
HEWL693975-901EA 30110 CZK
HEWL693975-901 HEWL699775-901 HEWL693775-901 HEWL699975-901 HEWL699975-301 HEWL695975-901 HEWL695975-301 HEWL695775-901 HEWL693975-301 HEWL699770-901 HEWL699770-901K HEWL693770-901K HEWL693970-301K HEWL693970-901K HEWL699970-901 HEWL699970-901K HEWL695770-901 HEWL695770-901K HEWL695970-301 HEWL695970-301K HEWL695970-901 HEWL695970-901K HEWL699970-301 HEWL699970-301K HEWL650750-901K HEWL690970-301K HEWL820750-926 HEWL821725-926 HEWL823750-926 HEWL690970-901K HEWL823750-940 HEWL820750-920 HEWL821725-919 HEWL820750-918 HEWL820750-919 HEWL821725-918 HEWL821725-920 HEWL823750-917 HEWL823750-918 HEWL823750-919 HEWL823750-920 HEWL820750-917 HEWL821725-917 HEWL695675-902 HEWL693675-912 HEWL693675-408 HEWL693675-901 HEWL693675-902 HEWL693675-906 HEWL695675-408 HEWL695675-901 HEWL695675-906 HEWL695675-912 HEWL821725-940 HEWL821725-941 HEWL821725-942 HEWL821725-943 HEWL821725-944
UHPLC columns, InfinityLab Poroshell 120
Kolony chromatografické
These InfinityLab Poroshell 120 1,9 µm columns provide high performance under the highest pressures.
  • Ultra-fast separations increase productivity
  • High resolution separations increase the accuracy and precision of analysis results, reducing the need for re-work
Stability for high-pH mobile phases
InfinityLab Poroshell HPH-C18 particles are made by chemically modifying the porous layer using proprietary technology that provides high-pH stability. That means you can use InfinityLab Poroshell 120 1,9 µm columns for all your fast LC method development needsMore Product Information