Tisk…
Bulk sorbents, SampliQ
Dodavatel: Agilent
5982-1082 5982-1382 5982-1882 5982-2082 5982-2282 5982-4382 5982-4482 5982-5752 5982-8082 5982-8382
HEWL5982-1082EA 3380 CZK
HEWL5982-1082 HEWL5982-1382 HEWL5982-1882 HEWL5982-2082 HEWL5982-2282 HEWL5982-4382 HEWL5982-4482 HEWL5982-5752 HEWL5982-8082 HEWL5982-8382
Bulk sorbents, SampliQ
Chromatografii, Sorbenty Flash chromatografické sorbenty
QuEChERS Bulk Salts and Sorbents make sample prep easy.
  • Prepackaged QuEChERS Kits are an easy way to capture the time-saving benefits of QuEChERS sample preparation
  • Extraction kits with preweighed anhydrous salts in sealed packets allow you to add salts after you add organic solvent to your sample – avoiding an exothermic reaction that can compromise analyte recovery
  • Dispersive kits with sorbents and salts supplied in 2 or 15 ml centrifuge tubes accommodate the aliquot volumes specified by current AOAC and EN methodologies
  • Universal dispersive kits provide excellent recoveries and reproducibility for all types of fruits and vegetables
  • Ceramic homogenizers break up salt agglomerates, promoting consistent sample extraction and increasing product recovery during extraction and dispersion; shaking time reduced from 60 to 20 seconds
If you prefer to pack your own tubes for QuEChERS, these bulk salts and sorbents provide high-quality materials for that purpose.