Tisk…
Single element standards for ICP-OES, calcium
Dodavatel: Agilent
5190-8329 5190-8330 5190-8368 5190-8369
HEWL5190-8329EA 2260 CZK
HEWL5190-8329 HEWL5190-8330 HEWL5190-8368 HEWL5190-8369
Single element standards for ICP-OES, calcium
Standardy Standardy kationické Standardy - vápník
They offer a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Informace o dodávce: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Balení: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.