Tisk…
Gold standard syringes for autosampler
Dodavatel: Agilent
5181-1267 5181-1273 5181-3360 5181-8810 5183-0316 5183-2058 5183-4729 5183-4730 9301-0713 9301-0725
548-0916EA 2230 CZK
548-0916 548-0917 HEWL5181-3360 HEWL5181-8810 548-0925 HEWL5183-2058 HEWL5183-4729 HEWL5183-4730 548-0926 HEWL9301-0725
Gold standard syringes for autosampler
Stříkačky Stříkačky chromatografické
These syringes with fixed needle are compatible with Agilent autosamplers 7693A and 7683.
* with PTFE-tipped plunger