Tisk…
Dissection set
Dodavatel: HERENZ HEINZ
1110401
HERE1110401EA 2940 CZK
HERE1110401
Dissection set
Sady řezací
This dissection set includes nine instruments in a strong wooden case.
Content:
1 scalpel handle No. 4 with two blades
1 scalpel
1 scalpel handle No. 3 with two blades
1 forceps
1 scissors (pointed)
2 dissection needles (1 pointed, 1 with blade)