Tisk…
Contact plates, RT/RTplus, single bagged
Dodavatel: MERCK LIFE SCIENCE
1462000020 1462530020 1465540020 1461890020 1461910020 1462660020 1464270020 1461540020 1465490020
HEIP1462000020EA 1670 CZK
HEIP1462000020 HEIP1462530020 HEIP1465540020 HEIP1461890020 HEIP1461910020 HEIP1462660020 HEIP1464270020 HEIP1461540020 HEIP1465490020
Contact plates, RT/RTplus, single bagged
Média mikrobiologická Misky mikrobiologické kontaktní
RT/RTplus contact plates are the ideal choice for sampling dry, sanitised surfaces in less critical areas. They are single-bagged and non irradiated.
  • Data matrix barcode on each plate for paperless and secure identification of individual plates
  • CLOSED and VENT closure options of RTplus contact plates for safe transportation and variation of incubation conditions
  • Convenient room temperature stability allows storage at site of use