Tisk…
Hei-CHILL 5000 Circulating Chillers
Dodavatel: Heidolph Instruments GmbH & Co.KG
591-01710-00
HEID591-01710-00EA 0 CZK
HEID591-01710-00
Hei-CHILL 5000 Circulating Chillers
Termostaty oběhové Termostaty oběhové Termostat cirkulační s chlazením
Comprehensive spectrum of services for demanding temperature control tasks with a cooling capacity of 5000 W.
  • Filling opening at the top, drain tap at the rear
  • Integrated programmer with 150 segments, can be divided into 5 programs
  • Electronic level indicator and low-level alarm
  • SmartCool system for digital, energy-saving cooling control, including automatic compressor control
  • Clearly arranged control elements, large TFT display and membrane keypad
  • USB interface and an alarm contact are integrated as standard features