Tisk…
Magnetic stirring bars set, cylindrical
Dodavatel: Heidolph Instruments GmbH & Co.KG
509-56000-00
HEID509-56000-00EA 736 CZK
HEID509-56000-00
Magnetic stirring bars set, cylindrical
Míchadla Míchadla magnetická
Set of PTFE covered magentic stirring bars.
  • Very good chemical resistance
  • Temperature resistant, suitable for high and low temperature applications
Set includes 1× each of stirrers with the following dimensions, L (mm): 25, 40 and 50 mm