Tisk…
Electronic Overhead Stirrers, Hei-TORQUE Platinum Package
Dodavatel: Heidolph Instruments GmbH & Co.KG
501-64029-04
HEID501-64029-04EA 102300 CZK
HEID501-64029-04
Electronic Overhead Stirrers, Hei-TORQUE Platinum Package
Míchadla Míchadla hřídelová
Hei-TORQUE Platinum package with the perfect end-to-end solution for powerful stirring and the simplest operation in the laboratory.
  • Hei-TORQUE Ultimate 400
  • Telescope stand
  • Clamp
Balení: Each package contains a telescopic stand and a suitable clamp for easy use on lab tables.