Tisk…
Beakers, with handle, porcelain
Dodavatel: GLASWARENFABRIK KARL HECHT
47020030 47020066 47020048 47020024 47020060
HECH47020030EA 797 CZK
HECH47020030 HECH47020066 HECH47020048 HECH47020024 HECH47020060
Beakers, with handle, porcelain
Kádinky
Porcelain, glazed white.
  • Exceptional chemical resistance
  • Excellent thermal resistance
  • Robust, heavy construction
Cylindrical, with spout. With very easy-to-read, internal graduations.