Tisk…
Hydrometer according to Richter and Tralles
Dodavatel: GLASWARENFABRIK KARL HECHT
46175010
HECH46175010EA 696 CZK
HECH46175010
Hydrometer according to Richter and Tralles
Hydrometry
Hydrometer designed without thermometer.
  • Reference temperature: 20 °C (68 °F)
Balení: In a protective plastic tube.