Tisk…
Stoppers for standard ground joints, octogonal, HDPE
Dodavatel: GLASWARENFABRIK KARL HECHT
42990007 42990010 42990012 42990014 42990019 42990024 42990029 42990034 42990045
HECH42990007EA 15.4 CZK
HECH42990007 HECH42990010 HECH42990012 HECH42990014 HECH42990019 HECH42990024 HECH42990029 HECH42990034 HECH42990045
Stoppers for standard ground joints, octogonal, HDPE
Zátky Zátky PE/LDPE/HDPE
High-density polyethylene.
  • Acid and alkali resistant
  • Octogonal