Tisk…
Clips for conical ground joints
Dodavatel: GLASWARENFABRIK KARL HECHT
42386010 42386012 42386034 42386019 42386024
HECH42386010EA 828 CZK
HECH42386010 HECH42386012 HECH42386034 HECH42386019 HECH42386024
Clips for conical ground joints
Svorky Svorky pro kónické NZ
Clips made of POM for conical ground joints.
  • Material does not scratch glass surfaces
  • Resistant to temperatures up to +150 °C