Tisk…
Volumetric flasks, class B, white markings
Dodavatel: GLASWARENFABRIK KARL HECHT
42089092
HECH42089092EA 2410 CZK
HECH42089092
Volumetric flasks, class B, white markings
Baňky Baňky odměrné
Borosilicate glass 3.3.
  • Higher accuracy than standard class B, Tolerance: 1,5 times of the official tolerance of class A
  • Calibrated to contain (marked 'In')
*Wide neck
Certifikace: In accordance with DIN EN ISO 1042