Tisk…
Blood sedimentation apparatus
Dodavatel: GLASWARENFABRIK KARL HECHT
40367010
HECH40367010EA 4970 CZK
HECH40367010
Blood sedimentation apparatus
Systémy pro klinickou diagnostiku Clinical Instruments
Blood sedimentation apparatus stand of stainless steel with tilting device for rapid sedimentation.
  • Pipettes with fused graduation 'ASSISTENT-PERMANENT'
Informace pro objednávání: Complete with blood sedimentation pipettes, blood mixing glasses with rubber stopper, special rubber caps (macro method only) and directions for use.
Balení: In a box.