Tisk…
Storage and transport boxes, DURAporter™
Dodavatel: HEATHROW
HS120052 HS120077 HS120078
HEATHS120052EA 5260 CZK
HEATHS120052 HEATHS120077 HEATHS120078
Storage and transport boxes, DURAporter™
Krabice Krabice přepravní
PC, transparent.
  • Silicone water-tight seal and three lid clasps provide secure closure
  • Lid opens a full 180°, enabling total access to contents and making it easier to clean
  • Will accommodate 13 and 16 mm tubes in a 72-place rack
  • In compliance with U.S. standards, OSHA 29 CFR 1910.1030 and meets UN3373 compliant transport unit requirements
The DURAporter™ is ideal for transporting biological and clinical samples, instruments and products that needs to be kept clean and dry under testing environmental conditions. The carry handle folds neatly into theMore Product Information