Tisk…
Stroje sítovací, Tyler Ro-Tap®
Dodavatel: VWR Collection
510-0945EA 130630 CZK
510-0945 510-0947 510-1035 510-1036 HAVE205322655 HAVE205322761
Stroje sítovací, Tyler Ro-Tap®
Třepačky Stroje sítovací
For dry sieving. Tyler Ro-Tap® shakers RX 29 CE and RX 30 CE reproduce the circular and tapping motion of hand sieving, but in a repeatable mechanical action. This sieve shaker makes a circular movement of the sieve stack and a mechanical arm makes a tapping movement on the top of the sieve stack, thus creating a perfect and repeatable imitation of hand shaking with additional tapping of the sieve on the side. They are operated in a totally enclosed cabinet, which guarantees the safety of the operator, and reduces the sound emissions significantly.
  • Rotation frequency: 278 min⁻¹;
More Product Information