Tisk…
Surgical masks, Foliodress® Mask Protect Senso
Dodavatel: HARTMANN PAUL
9925351
HART9925351EA 390 CZK
HART9925351
Surgical masks, Foliodress® Mask Protect Senso
Masky Roušky chirurgické
Comfortable surgical face masks made from special combination of particularly soft, lightweight and hypoallergenic non-woven materials.
  • For wearers with sensitive skin
  • Integrated filter for a high bacterial filtration efficiency and low breathing resistance
  • Long integrated nosepiece
  • Ultrasonically sealed ties
  • Free from natural rubber latex, fragrance free and durable
Suitable for use in all surgical areas, like isolation wards, intensive care departments, delivery rooms, premature and newborn infant wards. Ideal for outpatient use and emergency.