Tisk…
Flip tube A
Dodavatel: HAMILTON ROBOTICS
235693 235454
HAMR235693EA 15820 CZK
HAMR235693 HAMR235454
Flip tube A
Zkumavky Zkumavky robotické
The consumable flip tube® A is especially designed to automate the opening / closing of the lid by a controlled automated process, that applies a stepwise gentle pressure on the rear end of the hinged lid, enabling a fully walkaway automated process.
  • Made from highly transparent polypropylene for easy optical control
  • Contamination-free opening manually or by FlipTube® tool
  • Precise lid sealing to minimize sample evaporation
  • Pierceable lid and easy marking due to large frosted lid and surface on the side
  • Available in 'standard quality', 'biological