Tisk…
Conductivity cell, platinum, sensION+ 5060
Dodavatel: Hach
LZW5060970002
HACHLZW5060970002EA 13590 CZK
HACHLZW5060970002
Conductivity cell, platinum, sensION+ 5060
Elektrody Cely vodivostní
This three-pole platinum cell with integrated temperature sensor is protected against harsh field conditions.
  • Heavy-duty electrode handle design is optimised for field calibration and storage
  • Calibration and storage tube screws directly onto the heavy-duty handle
  • Resilient polycarbonate body and durable MP5 connector ensure protected, repeatable performance in the field
  • Dedicated for use with Hach sensION+ portable conductivity meters
The 5060 is ideal for conductivity measurements in general aqueous applications.
Informace o dodávce: Supplied with basic user manual.