Tisk…
Radiometer Analytical XR110 Reference Electrode (Red Rod, Ground Joint, Screw Cap)
Dodavatel: Hach
B20B112
HACHB20B112EA 10250 CZK
HACHB20B112
Radiometer Analytical XR110 Reference Electrode (Red Rod, Ground Joint, Screw Cap)
Elektrody Elektrody ostatní
The XR110 reference electrode has a glass body, red rod Ag/AgCl reference technology system, refillable electrolyte (KCl 3 M), ground joint, and porous pin junction.
  • Recommended for voltammetry applications and educational purposes
The XR110 is a screw cap electrode and requires an external connection cable for connection to a meter.
Informace o dodávce: Delivery includes radiometer analytical XR110 reference electrode, 30 ml KCl 3M refill solution and operating instructions.