Tisk…
Potassium chloride standard solution
Dodavatel: Hach
2714349 2974426 2974226 2974326
HACH2714349EA 2280 CZK
HACH2714349 HACH2974426 HACH2974226 HACH2974326
Potassium chloride standard solution
Standardy Standardy vodivostní
For calibrations/accuracy checks on conductivity/salinity meters.