Tisk…
BOD apparatus, BODTrak™II
Dodavatel: Hach
2952400
HACH2952400EA 242650 CZK
HACH2952400
BOD apparatus, BODTrak™II
Biologická spotřeba kyslíku (BOD) měřiče
The BODTrak™ II apparatus is easy to set up and allows for quick sample preparation for BOD analysis.
  • Standalone operation
  • Large graphic display that continuously updates results that can be displayed as a curve
  • Magnetic stirring of each bottle for more rapid results than dilution method
  • Selectable range and test duration
Informace o dodávce: Supplied with six bottles, six magnetic stir bars, six seal cups, spatula scoop, 50 BOD nutrient buffer pillows and potassium hydroxide pellets