Tisk…
Metals quality control standard for drinking water
Dodavatel: Hach
2833649 2833749
HACH2833649EA 1980 CZK
HACH2833649 HACH2833749
Metals quality control standard for drinking water
Standardy Standardy pro environmentální analýzu
These mixed standards are ready to use with no dilutions required. It include standards for determination of copper, iron and manganese.