Tisk…
Racks for test tubes
Dodavatel: Hach
2497902 1488600
HACH2497902EA 1250 CZK
HACH2497902 HACH1488600
Racks for test tubes
Stojany Tube Racks
These test tube racks features a special blend of polyethylene which sinks in water, assuring non-floating racks when immersed.
  • Stackable
  • Good chemical resistance
  • Good temperature resistance