Tisk…

3-Aminopicolinic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 3-Amino-2-pyridinecarboxylic acid, 3-Aminopyridine-2-carboxylic acid, 2-Pyridinecarboxylic acid, 3-amino-

H64861.06 H64861.14 H64861.22
H64861.06EA 1880 CZK
H64861.06 H64861.14 H64861.22
3-Aminopicolinic acid 97%
3-Aminopicolinic acid

Purity: 98% CAS No.: 1462-86-8 Molecular Formula: C6H6N2O2 Formula Weight: 138.13 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: Brown Melting Point: 218-222° Beilstein Reference: 3600 MDL No.: MFCD00090153

Vzorec: C₆H₆N₂O₂
MW: 138.126 g/mol
Bod tání: 218…222 °C
MDL Number: MFCD00090153
CAS číslo: 1462-86-8
EINECS: 215-971-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR