Tisk…

4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride 95%

Dodavatel: Thermo Fisher Scientific
Varování

H64755.MD H64755.03
H64755.MDEA 3680 CZK
H64755.MD H64755.03
4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride 95%
4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride

Purity: 95% CAS No.: 879668-17-4 Molecular Formula: C6H9N3.HCl Formula Weight: 159.62 Physical Form: Solid Beilstein Reference: 15312806 MDL No.: MFCD11846200

Vzorec: C₆H₁₀ClN₃
Storage Temperature: Ambient
MDL Number: MFCD11846200
CAS číslo: 879668-17-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR