Tisk…

5-Bromo-2-hydroxy-4-picoline 97%

Dodavatel: Thermo Fisher Scientific
Varování

H64674.06 H64674.14
H64674.06EA 1860 CZK
H64674.06 H64674.14
5-Bromo-2-hydroxy-4-picoline 97%
5-Bromo-2-hydroxy-4-picoline

Purity: 97% CAS No.: 164513-38-6 Molecular Formula: C6H6BrNO Formula Weight: 188.02 Physical Form: Solid Appearance: Off-white Melting Point: 198-202° Beilstein Reference: 6798531 MDL No.: MFCD03427664

Vzorec: C₆H₆BrNO
Bod tání: 198…202 °C
Storage Temperature: Ambient
MDL Number: MFCD03427664
CAS číslo: 164513-38-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR