Tisk…

5-Bromo-2-iodobenzoic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

H64643.06 H64643.14
H64643.06EA 1610 CZK
H64643.06 H64643.14
5-Bromo-2-iodobenzoic acid 97%
5-Bromo-2-iodobenzoic acid

Purity: 97% CAS No.: 21740-00-1 Molecular Formula: C7H4BrIO2 Formula Weight: 326.92 Physical Form: Solid Sensitivity: Light Sensitive Melting Point: 158-163° Beilstein Reference: 3247380 MDL No.: MFCD00079717

Vzorec: C₇H₄BrIO₂
MW: 326.92
Bod tání: 158…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00079717
CAS číslo: 21740-00-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR