Tisk…

2-Bromo-6-chlorobenzaldehyde 98%

Dodavatel: Thermo Fisher Scientific
Varování

H64605.03 H64605.06
H64605.03EA 3610 CZK
H64605.03 H64605.06
2-Bromo-6-chlorobenzaldehyde 98%
2-Bromo-6-chlorobenzaldehyde

Purity: 98% CAS No.: 64622-16-8 Molecular Formula: C7H4BrClO Formula Weight: 219.46 Physical Form: Solid Melting Point: 81-86° Beilstein Reference: 2613087 MDL No.: MFCD00266792

Vzorec: C₇H₄BrClO
Bod tání: 81…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00266792
CAS číslo: 64622-16-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR