Tisk…

4-Nitro-1H-indole-3-carbaldehyde 97%

Dodavatel: Thermo Fisher Scientific
Varování

H63595.03 H63595.06
H63595.03EA 3540 CZK
H63595.03 H63595.06
4-Nitro-1H-indole-3-carbaldehyde 97%
4-Nitro-1H-indole-3-carbaldehyde

Purity: 97% CAS No.: 10553-11-4 Molecular Formula: C9H6N2O3 Formula Weight: 190.16 Physical Form: Solid Appearance: Yellow Melting Point: 189-193° MDL No.: MFCD04973979

Vzorec: C₉H₆N₂O₃
Bod tání: 189…193 °C
Storage Temperature: Ambient
MDL Number: MFCD04973979
CAS číslo: 10553-11-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR