Tisk…

H-Lys(boc)-OtBu·HCl 95%

Dodavatel: Thermo Fisher Scientific

Synonyms: Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride, H-Lys(Boc)-OtBu.HCl, H-Lys(Boc)-OtBu·HCl

H62993.03 H62993.06
H62993.03EA 3920 CZK
H62993.03 H62993.06
H-Lys(boc)-OtBu·HCl 95%
H-Lys(boc)-OtBu·HCl

Purity: 95% CAS No.: 13288-57-8 Molecular Formula: C15H30N2O4.HCl Formula Weight: 338.87 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00038899

Vzorec: C₁₅H₃₁ClN₂O₄
MW: 338.873 g/mol
MDL Number: MFCD00038899
CAS číslo: 13288-57-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR