Tisk…

2-Fluoro-6-methoxybenzeneboronic acid pinacol ester 96%

Dodavatel: Thermo Fisher Scientific

H62812.MD H62812.03
H62812.MDEA 4260 CZK
H62812.MD H62812.03
2-Fluoro-6-methoxybenzeneboronic acid pinacol ester 96%
2-Fluoro-6-methoxybenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C13H18BFO3 Formula Weight: 252.09 Physical Form: Solid

Vzorec: C₁₃H₁₈BFO₃
Storage Temperature: Ambient
MDL Number: MFCD16996294
CAS číslo: 1599432-41-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR