Tisk…

3-Fluoro-2-nitrophenol ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H61670.03 H61670.06
H61670.03EA 1310 CZK
H61670.03 H61670.06
3-Fluoro-2-nitrophenol ≥97%
3-Fluoro-2-nitrophenol
Vzorec: C₆H₄FNO₃
Storage Temperature: Ambient
MDL Number: MFCD07368753
CAS číslo: 385-01-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR