Tisk…

Piperonyl butoxide tech. 90%, Technical Grade

Dodavatel: Thermo Fisher Scientific

Synonyms: 5-[2-(2-Butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole, 5-((2-(2-Butoxyethoxy)ethoxy)methyl)-6-propylbenzo[d][1,3]dioxole, NSC 8401

H60771.ACEA 783 CZK
H60771.AC H60771.AE
Piperonyl butoxide tech. 90%, Technical Grade
Piperonyl butoxide

Piperonyl butoxide is used as a pharmaceutical intermediate.


  • Soluble in benzene, ethanol and methanol

Vzorec: C₁₉H₃₀O₅
MW: 338.44 g/mol
Bod varu: 155 °C (0.3 mmHg)
Hustota: 1.059
Bod vzplanutí: 171 °C (340 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005842
CAS číslo: 51-03-6
EINECS: 200-076-7
UN: 3082
ADR: 9 III

Objednat

Výsledky specifikačního testu

Formula Weight 338,44
Purity 90%
Refractive Index 1,498
Assay (GC) >88,0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR