Tisk…

BMH (1,6-Bis(maleimido)hexane) 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: BMH

H58874.MCEA 4880 CZK
H58874.MC H58874.ME
BMH (1,6-Bis(maleimido)hexane) 97%
BMH (1,6-Bis(maleimido)hexane)

Purity: 97% CAS No.: 4856-87-5 Molecular Formula: C14H16N2O4 Formula Weight: 276.29 Storage Temperature: 2°C to 8°C Melting Point: 144° MDL No.: MFCD00047122

Vzorec: C₁₄H₁₆N₂O₄
MW: 276.29 g/mol
MDL Number: MFCD00047122
CAS číslo: 4856-87-5
EINECS: 225-449-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR