Tisk…

3-Pyridin-4-ylpropionic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

H58655.03 H58655.06
H58655.03EA 5080 CZK
H58655.03 H58655.06
3-Pyridin-4-ylpropionic acid 97%
3-Pyridin-4-ylpropionic acid

Purity: 97% CAS No.: 6318-43-0 Molecular Formula: C8H9NO2 Formula Weight: 151.16 Melting Point: 231° Beilstein Reference: 122234 MDL No.: MFCD00084846

Vzorec: C₈H₉NO₂
Bod tání: 231 °C
Storage Temperature: Ambient
MDL Number: MFCD00084846
CAS číslo: 6318-43-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR