Tisk…

4-Fluoro-2-iodobenzoic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H55531.03 H55531.06
H55531.03EA 1960 CZK
H55531.03 H55531.06
4-Fluoro-2-iodobenzoic acid ≥97%
4-Fluoro-2-iodobenzoic acid

Purity: 97% CAS No.: 56096-89-0 Molecular Formula: C7H4FIO2 Formula Weight: 266.01 Melting Point: 143-147° UN No.: 2811 MDL No.: MFCD04108278

Vzorec: C₇H₄FIO₂
MW: 266.01 g/mol
Bod tání: 143…147 °C
Storage Temperature: Ambient
MDL Number: MFCD04108278
CAS číslo: 56096-89-0
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR