Tisk…

4-Amino-2-methylpyridine 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H55036.MD H55036.03 H55036.06
H55036.MDEA 1600 CZK
H55036.MD H55036.03 H55036.06
4-Amino-2-methylpyridine 98%
4-Amino-2-methylpyridine

Purity: 98% CAS No.: 18437-58-6 Molecular Formula: C6H8N2 Formula Weight: 108.14 Melting Point: 93-97° MDL No.: MFCD00186509

Vzorec: C₆H₈N₂
Bod tání: 93…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00186509
CAS číslo: 18437-58-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR