Tisk…

2-Amino-4-chloropyrimidine 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H55007.03 H55007.06
H55007.03EA 2250 CZK
H55007.03 H55007.06
2-Amino-4-chloropyrimidine 98%
2-Amino-4-chloropyrimidine

Purity: 98% CAS No.: 3993-78-0 Molecular Formula: C4H4ClN3 Formula Weight: 129.55 Storage Temperature: 2°C to 8°C Melting Point: 155-160° dec. MDL No.: MFCD00038021

Vzorec: C₄H₄ClN₃
Storage Temperature: Refrigerator
MDL Number: MFCD00038021
CAS číslo: 3993-78-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR