Tisk…

(1S,3R)-(-)-Camphoric acid ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: (1S,3R)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid

H54042.MD H54042.03 H54042.06
H54042.MDEA 1310 CZK
H54042.MD H54042.03 H54042.06
(1S,3R)-(-)-Camphoric acid ≥98%
(1S,3R)-(-)-Camphoric acid

Purity: 98% CAS No.: 560-09-8 Molecular Formula: C10H16O4 Formula Weight: 200.23 Melting Point: 188-190° Beilstein Reference: 1874674 Merck Reference: 14,1733 Optical Rotation: -48° (c=10 in ethanol) MDL No.: MFCD00064937

Vzorec: C₁₀H₁₆O₄
MW: 200.23 g/mol
Bod tání: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00064937
CAS číslo: 560-09-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR