Tisk…

4-(N,N-Diphenylamino)phenylboronic acid 98%

Dodavatel: Thermo Fisher Scientific
Varování

H52902.MDEA 2120 CZK
H52902.MD
4-(N,N-Diphenylamino)phenylboronic acid 98%
4-(N,N-Diphenylamino)phenylboronic acid

Purity: 98% CAS No.: 201802-67-7 Molecular Formula: C18H16BNO2 Formula Weight: 289.14 Melting Point: 110-115° MDL No.: MFCD06798117

Vzorec: C₁₈H₁₆BNO₂
Bod tání: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD06798117
CAS číslo: 201802-67-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR