Tisk…

(R)-3-(Boc-amino)piperidine 97%

Dodavatel: Thermo Fisher Scientific
Varování

H52749.03 H52749.06
H52749.03EA 1730 CZK
H52749.03 H52749.06
(R)-3-(Boc-amino)piperidine 97%
(R)-3-(Boc-amino)piperidine

Purity: 97% CAS No.: 309956-78-3 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 MDL No.: MFCD03093382

Vzorec: C₁₀H₂₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03093382
CAS číslo: 309956-78-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR