Tisk…

(1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

H52747.MD H52747.03
H52747.MDEA 3780 CZK
H52747.MD H52747.03
(1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid 97%
(1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid

Purity: 97% CAS No.: 96443-42-4 Molecular Formula: C8H12O4 Formula Weight: 172.18 MDL No.: MFCD04972280

Vzorec: C₈H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD04972280
CAS číslo: 96443-42-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR