Tisk…

Ethyl-4-methyloxazole-5-carboxylate ≥97%

Dodavatel: Thermo Fisher Scientific

H52404.03 H52404.06
H52404.03EA 4500 CZK
H52404.03 H52404.06
Ethyl-4-methyloxazole-5-carboxylate ≥97%
Ethyl-4-methyloxazole-5-carboxylate

Purity: 97+% CAS No.: 20485-39-6 Molecular Formula: C6H7NO3 Formula Weight: 141.10 Physical Form: Liquid Appearance: Colourless MDL No.: MFCD00062573

Vzorec: C₇H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00062573
CAS číslo: 20485-39-6
EINECS: 243-848-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR