Tisk…

Boc-L-β-homoleucine 95%

Dodavatel: Thermo Fisher Scientific

Synonyms: Boc-β-HoLeu-OH, (S)-3-((tert-Butoxycarbonyl)amino)-5-methylhexanoic acid

H52173.MD H52173.03
H52173.MDEA 3990 CZK
H52173.MD H52173.03
Boc-L-β-homoleucine 95%
Boc-L-beta-homoleucine

Purity: 95% CAS No.: 132549-43-0 Molecular Formula: C12H23NO4 Formula Weight: 245.32 Melting Point: 53-55° Beilstein Reference: 4251252 MDL No.: MFCD02101665

Vzorec: C₁₂H₂₃NO₄
MW: 245.32 g/mol
Bod tání: 53…55 °C
Storage Temperature: Ambient
MDL Number: MFCD02101665
CAS číslo: 132549-43-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR