Tisk…

Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 95%

Dodavatel: Thermo Fisher Scientific

H52167.MD H52167.03
H52167.MDEA 2990 CZK
H52167.MD H52167.03
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 95%
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH

Purity: 95% CAS No.: 186320-06-9 Molecular Formula: C22H19NO4S Formula Weight: 393.46 Beilstein Reference: 9512976 MDL No.: MFCD00151916

Vzorec: C₂₂H₁₉NO₄S
Storage Temperature: Ambient
MDL Number: MFCD00151916
CAS číslo: 186320-06-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR