Tisk…

N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine 95%

Dodavatel: Thermo Fisher Scientific

H51988.03 H51988.06
H51988.03EA 3220 CZK
H51988.03 H51988.06
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine 95%
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine

Purity: 95% CAS No.: 114873-01-7 Molecular Formula: C14H18FNO4 Formula Weight: 283.30 Melting Point: 75-80° Beilstein Reference: 5347375 Optical Rotation: +6.5° (c=1 in methanol) MDL No.: MFCD00672522

Vzorec: C₁₄H₁₈FNO₄
Bod tání: 75…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00672522
CAS číslo: 114873-01-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR